1,4-Bis(diphenylphosphino)butan

Lieferant: Molekula

Synonyms: Butan-1,3-diylbis(diphenylphosphin)Butylenbis(diphenylphosphin)Bis(1,2-diphenylphosphino)butan

MOLEM82971615EA 0 CHF
MOLEM82971615
1,4-Bis(diphenylphosphino)butan
1,4-Bis(diphenylphosphino)butan
Formel: C₂₈H₂₈P₂
MW: 426,48 g/mol
Melting Pt: 195 °C
Dichte: 1,23 g/cm³ (20 °C)
MDL Number: MFCD00003051
CAS-Nummer: 7688-25-7
EINECS: 231-698-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR