1,3-Bis(diphenylphosphino)propan
Lieferant: Molekula
|
Achtung
|
Synonyms:
Bis(1,2-diphenylphosphino)propan
, Propylenbis(diphenylphosphin)
, Propan-1,3-diylbis(diphenylphosphin)
MOLE28001437-5GEA
0
CHF
MOLE28001437-5G
1,3-Bis(diphenylphosphino)propan
1,3-Bis(diphenylphosphino)propan
Formel:
C₂₇H₂₆P₂ MW: 412,45 g/mol Melting Pt: 60…63 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003050 CAS-Nummer: 6737-42-4 EINECS: 229-791-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...