Seesand

Lieferant: Roth Carl

ROTH4309.1EA 142.35 CHF
ROTH4309.1
Seesand
Seesand
Formel: SiO₂
MW: 60,08 g/mol
Siedepunkt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Dichte: 2,66 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
CAS-Nummer: 7631-86-9
EINECS: 231-545-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR