Phenothiazin

Lieferant: LGC Standards PROMOCHEM
Achtung

PROOMM0157.02EA 765 CHF
PROOMM0157.02
Phenothiazin
Phenothiazin
Formel: C₁₂H₉NS
MW: 199,28 g/mol
Siedepunkt: 371 °C (1013 hPa)
Melting Pt: 182…185 °C
Dichte: 1,34 g/cm³ (20 °C)
Flash-Pt: 202 °C
MDL Number: MFCD00005015
CAS-Nummer: 92-84-2
EINECS: 202-196-5
Merck Index: 12,07404

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR