4-Aminoantipyrin
Lieferant: PanReac AppliChem
|
Achtung
|
Synonyms:
4-Aminophenazon
, 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-on
A1523.0100EA
221
CHF
A1523.0100
4-Aminoantipyrin
4-Aminoantipyrin
Formel:
C₁₁H₁₃N₃O MW: 203,24 g/mol Melting Pt: 107…109 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003145 CAS-Nummer: 83-07-8 EINECS: 201-452-3 Merck Index: 13,00596 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...