Penicillin G Kaliumsalz
Lieferant: EUROCLONE
|
Achtung
|
Synonyms:
6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt
EMR185010
EMR185025
EMR185100
EURLEMR185010EA
0
CHF
EURLEMR185010
EURLEMR185025
EURLEMR185100
Penicillin G Kaliumsalz
Penicillin G Kaliumsalz
Formel:
C₁₆H₁₇KN₂O₄S MW: 372.49 g/mol Melting Pt: 214…217 °C Dichte: 1.359 g/cm³ (20 °C) Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00036193 CAS-Nummer: 113-98-4 EINECS: 204-038-0 Merck Index: 12,07225 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...