γ-(N-Morpholino)propansulfonsäure (MOPS)

Lieferant: GERBU BIOCHEMICALS

Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

1081.0100 1081.0250
GRBU1081.0100EA 0 CHF
GRBU1081.0100 GRBU1081.0250
γ-(N-Morpholino)propansulfonsäure (MOPS)
γ-(N-Morpholino)propansulfonsäure (MOPS)
Formel: C₇H₁₅NO₄S
Melting Pt: > 250 °C
Flash-Pt: 110 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006183
CAS-Nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR