γ-(N-Morpholino)propansulfonsäure (MOPS)
Lieferant: EUROCLONE
EMR176100
EMR176500
EURLEMR176100EA
0
CHF
EURLEMR176100
EURLEMR176500
γ-(N-Morpholino)propansulfonsäure (MOPS)
γ-(N-Morpholino)propansulfonsäure (MOPS)
Formel:
C₇H₁₅NO₄S Melting Pt: > 250 °C Flash-Pt: 110 °C (closed cup) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00006183 CAS-Nummer: 1132-61-2 EINECS: 214-478-5 Merck Index: 13,06288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...