2-Ethylhexylacetat 99%, rein
Lieferant: Thermo Scientific
|
Achtung
|
Synonyms:
Essigsäure(2-ethylhexyl)ester
, (2-Ethylhexyl)acetat
ACRO410105000EA
62.2
CHF
ACRO410105000
2-Ethylhexylacetat 99%, rein
2-Ethylhexylacetat
Formel:
C₁₀H₂₀O₂ MW: 172,27 g/mol Siedepunkt: 197 °C Melting Pt: –45 °C Dichte: 0,87 g/cm³ Flash-Pt: 71 °C |
MDL Number:
MFCD00027249 CAS-Nummer: 103-09-3 EINECS: 203-079-1 Merck Index: 12,06860 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4210 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...