L(-)-Cystin
Lieferant: Molekula
Synonyms:
(2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure)
, (R)-(-)-Cystin
, (H-Cys-OH)2
MOLEM12297455EA
0
CHF
MOLEM12297455
L(-)-Cystin
L(-)-Cystin
Formel:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH MW: 240,3 g/mol Siedepunkt: 428 °C (1013 hPa) Melting Pt: 261…262 °C Dichte: 1,677 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00064228 CAS-Nummer: 56-89-3 EINECS: 200-296-3 Merck Index: 13,02811 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...