L(-)-Cystin

Lieferant: BACHEM BIOCHEMICA

Synonyms: (2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure)(R)-(-)-Cystin(H-Cys-OH)2

BACE4030444.0100EA 0 CHF
BACE4030444.0100 BACE4030444.1000 BACE4030444.0500
L(-)-Cystin
L(-)-Cystin
Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240,3 g/mol
Siedepunkt: 428 °C (1013 hPa)
Melting Pt: 261…262 °C
Dichte: 1,677 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064228
CAS-Nummer: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR