Calciumacetat xHydrat

Lieferant: EUROCLONE

Synonyms: Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate

EMR106001 EMR106500
EURLEMR106001EA 0 CHF
EURLEMR106001 EURLEMR106500
Calciumacetat xHydrat
Calciumacetat xHydrat
Formel: (H₃CCOO)₂Ca·nH₂O
MW: 176,18 g/mol
Dichte: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00150019
CAS-Nummer: 114460-21-8
EINECS: 200-540-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR