G418 Disulfat
Lieferant: EUROCLONE
|
Achtung
|
Synonyms:
G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
EMR260001
EMR260250
EMR260500
BIOZEMR260001EA
0
CHF
BIOZEMR260001
EURLEMR260250
EURLEMR260500
G418 Disulfat
G418 disulphate
Formel:
C₂₀H₄₄N₄O₁₈S₂ MW: 692.72 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00058314 CAS-Nummer: 108321-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...