Safety spectacles, Contemporary 572 Diamond, small
Lieferant: Univet
Contemporary Diamond
111-2459EA 33.7 CHF
111-2459
Safety spectacles, Contemporary 572 Diamond, small
Schutzbrillen
Contemporary series combines elegance and style in proetction, changing the concept of safety glasses.
  • Light and extremely tough
  • Integrated side shields
  • PA12 nylon frame with zero metallic components
  • Glossy varnish with shaded effect
  • Clear KN lenses, with anti-fog and anti-scratch ultra performing Vanguard PLUS treatment
Elegant and graceful for women, dynamic for men. Glasses can be employed for prolonged use as they do not create distorsions and allow a correct color perception, sensibly reducing visual fatigue.
Suitable for mechanical work in good visibility conditions.
Size: 52
Zertifizierungen: EN-166, EN-170.