(9-Anthracenylmethoxy)acetic acid ≥98,0% (durch HPLC) für Massenspektrometrie, Supelco®
Lieferant: Merck
|
Achtung
|
09131-100MGEA
425
CHF
09131-100MG
(9-Anthracenylmethoxy)acetic acid ≥98,0% (durch HPLC) für Massenspektrometrie, Supelco®
(9-Anthracenylmethoxy)acetic acid
Formel:
C₁₇H₁₄O₃ |
CAS-Nummer:
207408-91-1 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance (Color) | White to Dark Yellow |
Appearance (Form) | Powder, Crystals and or Chunk(s) |
Purity (HPLC Area %) | ≥ 98.0 % |
Carbon Content | 75.1 - 78.2 % |
Hydrogen Content | 5.1 - 5.5 % |
Infrared Spectrum | Conforms To Structure |
Recommended Retest Period | 35 Months |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...