Amberlite® IRN-150 hydrogen and hydroxide form Ionenaustauscherharz, nukleare Qualität, Sigma-Aldrich®

Lieferant: Merck

00234-1KGEA 353 CHF
00234-1KG
Amberlite® IRN-150 hydrogen and hydroxide form Ionenaustauscherharz, nukleare Qualität, Sigma-Aldrich®
Amberlite® IRN-150

Mixed bed resin for primary water chemistry control in once-through systems. Useful in industrial water treatment where as-supplied resin must have absolute minimum of ionic and non-ionic contamination.

MDL Number: MFCD00145822
CAS-Nummer: 79956-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR