Amberlite® IRA-410 (Cl) Ionenaustauscherharz, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
216569-500GEA
115
CHF
216569-500G
216569-100G
Amberlite® IRA-410 (Cl) Ionenaustauscherharz, Supelco®
Amberlite® IRA-410 (Cl)
Amberlite® IRA-410 was used as packing material for columns. Amberlite® IRA-410 was also used in urinary analysis by HPLC.
Strongly basic (type II) anion exchange resin for water conditioning, removing iodine ions, neutralization of solutions, capturing xanthan and xanthene dyes, etc.; better regeneration efficiency than type I base resin.
MDL Number:
MFCD00132710 CAS-Nummer: 9002-26-0 |