RALA320550
Lieferant: RAL (REACTIFS RAL SA)
720-0331EA 184 CHF
720-0331
RALA320550
Haemalum-Shorr staining combines a nuclear stain, Mayer Haemalum with a cytoplasmic stain, Shorr Solution. It is used to carry out the morphological study of spermatozoa (spermocytogram).
  • For P.A.S. Staining
Haemalum-Shorr staining combines a nuclear stain, Mayer Haemalum with a cytoplasmic stain, Shorr Solution. It is used to carry out the morphological study of spermatozoa (spermocytogram).
For P.A.S. STAINING