Human native kallikrein (from plasma)
Lieferant: Merck
Calbiochem®
420307-50EA 818 CHF
420307-50
Human native kallikrein (from plasma)
Proteine und Peptide
Native kallikrein from human plasma. A serine protease that releases bradykinin from kininogen. Present in the plasma as an inactive precursor, prokallikrein.