pH/Conductivity - Clay Soil, certified reference material, Supelco®
Lieferant: Merck
CRM498-100GEA 207 CHF
CRM498-100G
pH/Conductivity - Clay Soil, certified reference material, Supelco®
Standards Leitfähigkeits-Standards
Values of analytes vary lot to lot.
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.