Tetrahydroxyquinone dihydrate Indikator für Titrationen (Ba, SO4), Fluka™

Lieferant: Honeywell Chemicals
Achtung

87510-5GEA 401.12 CHF
87510-5G
Tetrahydroxyquinone dihydrate Indikator für Titrationen (Ba, SO4), Fluka™
Tetrahydroxyquinone dihydrate
Formel: C₆H₄O₆·₂H₂O
MW: 208,12 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001597
CAS-Nummer: 5676-48-2

Order Now

Spezifikation Testergebnisse

Ignition Residue ≤0.5%
Identity (IR) complying
Assay (calculated to the dried substance) 98.5 - 101.5 %
Colour of the solution complying
Clearness of the solution complying
Residue on ignition Max. 0.5 %
Application test complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR