Mop frame, Clino® CR
Lieferant: Pfennig Reinigungstechnik
Clino®
PPSA3500921EA 150 CHF
PPSA3500921
Mop frame, Clino® CR
Mopps
The clino synchro series mop frames, which were especially designed for the use in cleanrooms, were developed in accordance with the hygienic design guidelines and fulfil all requirements for the use in hygienically sensitive areas.
  • Very easy to keep clean
  • Contactless operation with MopDrop CR
  • With quick-change system ClinoConnect lock
  • No slipping of the mop
The innovative opening mechanism in connection with the minimised weight enables an extremely comfortable handling and the mop is no longer pulled underneath the frame.
Weight: 370 g