Ice flake machine
Lieferant: Hoshizaki
HOSHFM-170AKEEA 0 CHF
HOSHFM-170AKE
Ice flake machine
Eisbereiter
The FM-170AKE (-N) is a modular ice machine that, depending on the model, produces flake ice or nugget ice.
  • Computer controlled for optimum performance manufacturing process
  • Easy-to-clean air filter
The bearings and the evaporator are made of high quality material steel and that increases the lifespan of the parts, making the machine lasts longer and less often requires expensive maintenance.