(-)-Perillyl alcohol ≥98%, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

77311-1MLEA 99.5 CHF
77311-1ML
(-)-Perillyl alcohol ≥98%, Supelco®
(-)-Perillyl alcohol
Formel: C₁₀H₁₆O
MW: 152,24 g/mol
MDL Number: MFCD00062995
CAS-Nummer: 18457-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR