DNA transfection, CaPo transfection kit
Lieferant: OZ Biosciences
OZBICP90000EA 0 CHF
OZBICP90000
DNA transfection, CaPo transfection kit
Transfektionsreagenzien
CaPo transfection kit is rapid, simple and easy to use.
  • Ideal for virus production
  • High HEK 293 cells transfection efficiency
  • Suitable for producing recombinant proteins
  • Serum compatible
It reaches between 95 to 100% of HEK 293 transfected cells and a very high titer for virus production.
Sufficient for 100 assays in 100 mm culture dishes with 1 µg of DNA or 500 assays in 6-well plates with 4 µg of DNA.
Lieferumfang: Supplied with 1X HEPES buffered saline (4×15 ml) and 2,5 M CaCl₂ (3,5 ml).
Bitte beachten Store at –20 °C.