Antibody package, Protein G
Lieferant: Cytiva
29-0588-06EA 197 CHF
29-0588-06
Antibody package, Protein G
Chromatographiesäulen
This Protein G package enables convenient purification of most species of antibodies, including rat.
The HiTrap™ Protein G HP column enclosed is recommended for most species of antibodies, including rat. A HiTrap™ Desalting column is also provided for quick pH neutralisation of the eluted antibodies.
Lieferumfang: Delivery includes: HiTrap™ Protein G HP (1×1 ml) HiTrap™ Desalting (1×5 ml).