DNA transfection reagent, COSFect™
Lieferant: OZ Biosciences
COSFect™
OZBICF12500EA 0 CHF
OZBICF12500 OZBICF11000 OZBICF10500
DNA transfection reagent, COSFect™
Transfektionsreagenzien
COSFect™ is a high efficiency transfection reagent specifically developed for COS cell lines. It is a lipid-based reagent based on Tee-Technology (Triggered Endosomal Escape). The cationic design of the reagent allows high nucleic acid compaction for efficient transport into COS cells. The reagent is composed of lipids, leading to high viability, and is ready to use.
  • Highly efficient with COS cell lines
  • Ready to use, no need for additional buffer
  • Low nucleic acid amount minimises toxicity
  • High level of nucleic acid compaction
  • Easy and straightforward protocol
  • Compatible with any culture medium, medium change not required
Store at –20 °C.
Lieferumfang: Shipped at room temperature.