Western-Re-Probe™ reagent, Calbiochem®
Lieferant: Merck
Calbiochem®
WB59-100EA 506 CHF
WB59-100
Western-Re-Probe™ reagent, Calbiochem®
Elektrophorese-Reagenzien Western Blotting Reagents
This Western-Re-Probe™ reagent is validated for use in immunoblotting.
  • Immunoblotting analysed repeatedly with multiple probes
  • Saving on hard to get reagents and samples
  • Correction and reanalysis of unsatisfactory Western blots
  • Saving time spent in gel rerunning and transfer processes
Designed to dissociate and remove antibodies from membrane-bound proteins without destroying antigenic binding capacity. Recommended for stripping chemiluminescent or radioisotopic signals from blots. Not recommended for stripping colour-producing Eestern blots.
Supplied as a 5X solution sufficient for stripping 25 to 30 standard blots.
Lieferumfang: Supplied as a 5X solution sufficient for stripping 25 to 30 standard blots.
Bitte beachten Western-Re-Probe™ reagent can be used for stripping and re-probing membranes 2-3 times. Some loss of signal may be noticed after second cycle of stripping and re-probing.