2-Chlorpyrimidin

Lieferant: Molekula
Achtung

MOLE16639234-25GEA 0 CHF
MOLE16639234-25G
2-Chlorpyrimidin
2-Chloropyrimidine
Formel: C₄H₃ClN₂
MW: 114,53 g/mol
Siedepunkt: 199 °C (1013 hPa)
Melting Pt: 65…66 °C
MDL Number: MFCD00006060
CAS-Nummer: 1722-12-9
EINECS: 217-020-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR