6-Methyl-2-pyridon 97%

Lieferant: Thermo Scientific

Synonyms: 2-Hydroxy-6-methylpyridin

ACRO156390250EA 294 CHF
ACRO156390250 ACRO156390050
6-Methyl-2-pyridon 97%
6-Methyl-2-pyridon
Formel: C₆H₇NO
MW: 109,13 g/mol
Siedepunkt: 283…284 °C (1013 hPa)
Melting Pt: 155…160 °C
MDL Number: MFCD00006279
CAS-Nummer: 3279-76-3
EINECS: 221-919-5

Order Now

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR