2,3-Pyridindicarbonsäure
Lieferant: Chemicon
CHMCAB5916EA
1500
CHF
CHMCAB5916
2,3-Pyridindicarbonsäure
2,3-Pyridindicarbonsäure
Formel:
C₇H₅NO₄ MW: 167,12 g/mol Siedepunkt: 357 °C (1013 hPa) Melting Pt: 188…190 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00006295 CAS-Nummer: 89-00-9 EINECS: 201-874-8 Merck Index: 12,08258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...