7-Methylxanthin
Lieferant: LGC Standards PROMOCHEM
Synonyms:
3,7-Dihydro-7-methyl-1H-purin-2,6-dion
, 7-Methyl-1H,3H-purin-2,6-dion
, 7-Methyl-3,7-dihydro-1H-purin-2,6-dion
, 2,6-Dihydroxy-7-methylpurin
, 7-Methyl-2,6(1H,3H)-purindion
, 7-Methyl-3,7-dihydro-2H-purin-2,6-dion
PROOMM0176.05EA
1700
CHF
PROOMM0176.05
7-Methylxanthin
7-Methylxanthin
Formel:
C₆H₆N₄O₂ MW: 166,14 g/mol |
MDL Number:
MFCD00037979 CAS-Nummer: 552-62-5 EINECS: 209-019-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...