2-Amino-6-chlorpurin

Lieferant: Molekula

Synonyms: 6-Chlor-7H-purin-2-ylamin

MOLEM52216629EA 0 CHF
MOLEM52216629
2-Amino-6-chlorpurin
2-Amino-6-chlorpurin
Formel: C₅H₄ClN₅
MW: 169,57 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00075252
CAS-Nummer: 10310-21-1
EINECS: 233-686-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR