Cer(IV)oxid 2oz (57g) Polierpaste

Lieferant: Thermo Scientific
Achtung

Synonyms: Cerdioxyd

39802.LAEA 153 CHF
39802.LA
Cer(IV)oxid 2oz (57g) Polierpaste
Cer(IV)oxid

Polishing compound for IR crystal windows

Formel: CeO₂
MW: 172,11 g/mol
Siedepunkt: 3468 °C (1013 hPa)
Melting Pt: 2000 °C
Dichte: 7,13 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010933
CAS-Nummer: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR