Dimethylsulfoxid
Lieferant: EUROCLONE
Synonyms:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
EMR031001
EMR031100
EMR031500
EURLEMR031001EA
0
CHF
EURLEMR031001
EURLEMR031100
EURLEMR031500
Dimethylsulfoxid
Dimethylsulfoxid
Formel:
(CH₃)₂SO MW: 78.14 g/mol Siedepunkt: 189 Melting Pt: 18.5 Dichte: 1.101 g/cm³ (20 °C) Flash-Pt: 87 Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00002089 CAS-Nummer: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...