Ammoniumsulfat, Millipore®
Lieferant: Merck
Synonyms:
Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
M-Clarity™ quality level = MQ100
1.01211.1000
1.01211.5000
1.01211.9050
1.01211.1000EA
77.2
CHF
1.01211.1000
1.01211.5000
1.01211.9050
Ammoniumsulfat, Millipore®
Ammoniumsulfat
Formel:
(NH₄)₂SO₄ Melting Pt: 336…339 °C Dichte: 1.78 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |