Ammoniumsulfat

Lieferant: EUROCLONE

EMR092000 EMR092001 EMR092500
EURLEMR092000EA 0 CHF
EURLEMR092000 EURLEMR092001 EURLEMR092500
Ammoniumsulfat
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Dichte: 1,78 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR