Ammoniumperoxodisulfat (APS)

Lieferant: NATIONAL DIAGNOSTICS
Gefahr

EC-504-100G EC-504-25G
NADIEC-504-100GEA 0 CHF
NADIEC-504-100G NADIEC-504-25G
Ammoniumperoxodisulfat (APS)
Ammoniumperoxodisulfat (APS)
Formel: (NH₄)₂S₂O₈
Melting Pt: 120 °C
Dichte: 1.98 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003390
CAS-Nummer: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR