Ammoniumnitrat
Lieferant: EUROCLONE
|
Achtung
|
Synonyms:
Ammonsalpeter
EURLEMR281001EA
129
CHF
EURLEMR281001
EURLEMR281005
EURLEMR281500
Ammoniumnitrat
Ammoniumnitrat
Formel:
NH₄NO₃ MW: 80,04 g/mol Siedepunkt: 230 °C (1013 hPa) Melting Pt: 169 °C Dichte: 1,73 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00011425 CAS-Nummer: 6484-52-2 EINECS: 229-347-8 UN: 1942 ADR: 5.1,III Merck Index: 13,00538 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...