Ammoniumchlorid

Lieferant: EUROCLONE
Achtung

Synonyms: Ammonium muriate, Salmiac

EMR089001 EMR089005 EMR089500
EURLEMR089001EA 0 CHF
EURLEMR089001 EURLEMR089005 EURLEMR089500
Ammoniumchlorid
Ammoniumchlorid
Formel: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335
Dichte: 1.5256 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011420
CAS-Nummer: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR