Triphenylphosphin

Lieferant: Molekula
Achtung

Synonyms: Phosphortriphenyl

MOLE23253991-1KGEA 0 CHF
MOLE23253991-1KG
Triphenylphosphin
Triphenylphosphin
Formel: C₁₈H₁₅P
MW: 262,29 g/mol
Siedepunkt: 377 °C (1013 hPa)
Melting Pt: 79 °C
Dichte: 1,194 g/cm³ (20 °C)
Flash-Pt: 180 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003043
CAS-Nummer: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR