Tetramethylammoniumiodid

Lieferant: Molekula

MOLE26902676-500GEA 0 CHF
MOLE26902676-500G
Tetramethylammoniumiodid
Tetramethylammoniumiodid
Formel: C₄H₁₂IN
MW: 201,05 g/mol
Melting Pt: 300 °C
Dichte: 1,84 g/cm³ (20 °C)
MDL Number: MFCD00011629
CAS-Nummer: 75-58-1
EINECS: 200-881-3
UN: 2811
ADR: 6.1,III
Merck Index: 12,09364

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR