Acetylcholinchlorid
Lieferant: Molekula
Synonyms:
(2-Acetoxyethyl)trimethylammoniumchlorid
, Acetylcholiniumchlorid
MOLE29010529-100GEA
0
CHF
MOLE29010529-100G
Acetylcholinchlorid
Acetylcholinchlorid
Formel:
H₃CC(O)O(CH₂)₂N(CH₃)₃Cl MW: 181,66 g/mol Melting Pt: 151 °C Dichte: 1,263 g/cm³ (20 °C) |
MDL Number:
MFCD00011698 CAS-Nummer: 60-31-1 EINECS: 200-468-8 Merck Index: 13,00089 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...