Chlormequatchlorid

Lieferant: EHRENSTORFER GMBH
Achtung

Synonyms: Chlorcholinchlorid(2-Chlorethyl)trimethylammoniumchlorid2-Chlorethyltrimethylammoniumchlorid

EHERCA11340000EA 114 CHF
EHERCA11340000
Chlormequatchlorid
Chlormequatchlorid
Formel: ClCH₂CH₂N(CH₃)₃Cl
MW: 158,07 g/mol
Melting Pt: 243 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011869
CAS-Nummer: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,02120

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR