Tetrabutylphosphoniumbromid

Lieferant: Molekula

MOLE75425740-100GEA 0 CHF
MOLE75425740-100G
Tetrabutylphosphoniumbromid
Tetrabutylphosphoniumbromid
Formel: C₁₆H₃₆BrP
MW: 339,34 g/mol
Melting Pt: 95…101 °C
MDL Number: MFCD00011853
CAS-Nummer: 3115-68-2
EINECS: 221-487-8
UN: 3464
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR