Cetrimoniumbromid

Lieferant: EUROCLONE
Gefahr

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

EMR313100 EMR313500
EURLEMR313100EA 0 CHF
EURLEMR313100 EURLEMR313500
Cetrimoniumbromid
Cetrimoniumbromid
Formel: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364,45 g/mol
Melting Pt: 237…243 °C
Flash-Pt: 244 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011772
CAS-Nummer: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR