Natriumcarbonat
Lieferant: EUROCLONE
|
Achtung
|
Synonyms:
Calcined soda, Carbonic acid disodium salt
EMR210000
EMR210001
EMR210005
EMR210500
EURLEMR210000EA
0
CHF
EURLEMR210000
EURLEMR210001
EURLEMR210005
EURLEMR210500
Natriumcarbonat
Natriumcarbonat
Formel:
Na₂CO₃ Siedepunkt: 1600 °C (1013 hPa) Melting Pt: 854 °C Dichte: 2.53 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003494 CAS-Nummer: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,08596 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...