Fluorescein

Lieferant: Chemicon

Synonyms: 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoesäureResorcinphthalein2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)-benzoesäure9-(2-Carboxyphenyl)-6-hydroxy-(3H)-3-xanthenonFluoreszein6-Hydroxy-9-(2-carboxyphenyl)-(3H)-xanthen-3-on3′,6′-Dihydroxyspiro[2-benzofuran-3,9′-xanthen]-1-on6-Hydroxy-9-(2-carboxyphenyl)-(3H)-3-xanthenon

CHMCCBL97EA 0 CHF
CHMCCBL97
Fluorescein
Fluorescein
Formel: C₂₀H₁₂O₅
MW: 332,31 g/mol
Melting Pt: 314…316 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005050
CAS-Nummer: 2321-07-5
EINECS: 219-031-8
Merck Index: 13,04185
https://ch.vwr.com/stibo/web/std.lang.all/00/30/26660030.jpg https://ch.vwr.com/stibo/web/std.lang.all/57/40/2375740.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR