3,4,4',5-Tetrachlorbiphenyl (PCB Nr. 81)
Lieferant: EHRENSTORFER GMBH
EHERC20008100EA
147
CHF
EHERC20008100
3,4,4',5-Tetrachlorbiphenyl (PCB Nr. 81)
3,4,4',5-Tetrachlorbiphenyl (PCB Nr. 81)
Formel:
C₁₂H₆Cl₄ MW: 291,99 g/mol |
CAS-Nummer:
70362-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...