3,4,4',5-Tetrachlorbiphenyl (PCB Nr. 81)

Lieferant: LGC Standards PROMOCHEM

PROOCIL-PCB-81-CEA 1250 CHF
PROOCIL-PCB-81-C
3,4,4',5-Tetrachlorbiphenyl (PCB Nr. 81)
3,4,4',5-Tetrachlorbiphenyl (PCB Nr. 81)
Formel: C₁₂H₆Cl₄
MW: 291,99 g/mol
CAS-Nummer: 70362-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR